• USA Home
  • G4630 - Guanidine hydrochloride

EMAIL THIS PAGE TO A FRIEND
G4630 Sigma

Guanidine hydrochloride

≥98%

Synonym: Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidinium chloride

Purchase

Properties

Related Categories Biochemicals and Reagents, Chaotropic Agents, Core Bioreagents, Denaturation, Nucleosides and Nucleotides,
InChI Key   PJJJBBJSCAKJQF-UHFFFAOYSA-N
assay   ≥98%
contains   ≤0.4% SiO2 as anticaking agent
pH   (25 °C, 4.6 - 6/573 g/L)
mp   180-185 °C(lit.)
solubility   H2O: soluble573 g/L at 20 °C (completely)

Description

Application

Strong chaotropic agent useful for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary.

Other Notes

This product has been replaced by 50950-SIGMA | Guanidine hydrochloride ≥98%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
MF4300000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?