• USA Home
  • G4633 - Globulins Cohn fraction III bovine

EMAIL THIS PAGE TO A FRIEND
G4633 Sigma

Globulins Cohn fraction III bovine

lyophilized powder

Purchase

Properties

form   lyophilized powder
composition   Protein, ~75% biuret
storage temp.   2-8°C

Description

Other Notes

Predominantly γ- and β-globulins. Contains plasminogen activity.

Physical form

Lyophilized powder containing NaCl

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?