• USA Home
  • G4658 - α-D-Glucosamine pentaacetate

EMAIL THIS PAGE TO A FRIEND
G4658 Sigma

α-D-Glucosamine pentaacetate

Synonym: 2-Acetamido-2-deoxy-1,3,4,6-tetra-O-acetyl-α-D-glucopyranose

Purchase

Properties

storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?