EMAIL THIS PAGE TO A FRIEND
G4670 Sigma

Glufosinate Ammonium

Purchase

Properties

Featured Industry   Agriculture

Description

General description

Glufosinate Ammonium is a natural compound isolated from two species of Streptomyces fungi. It inhibits the activity of an enzyme, glutamine synthetase, which is necessary for the production of glutamine and for ammonia detoxification. The application of glufosinate ammonium leads to reduced glutamine and increased ammonia levels in the plant tissues. This causes photosynthesis to stop and the plant dies within a few days.

Glufosinate Ammonium inhibits the activity of glutamine synthetase, an enzyme necessary for photosynthesis.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
EK7713600
Flash Point(F) 
210 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?