• USA Home
  • G4783 - Glycerol Dehydrogenase from Enterobacter aerogenes

EMAIL THIS PAGE TO A FRIEND
G4783 Sigma

Glycerol Dehydrogenase from Enterobacter aerogenes

lyophilized powder, 20-80 units/mg protein, pH 10.0

Synonym: Glycerol:NAD+ 2-oxidoreductase

Purchase

Description

Unit Definition

One unit will oxidize 1.0 μmole of glycerol to dihydroxyacetone per min at 25 °C at pH 10.0.

Physical form

Lyophilized powder containing potassium phosphate buffer salts

Application

Glycerol dehydrogenase was used in the kinetic enzymatic determination of glycerol in wine and beer.

Biochem/physiol Actions

Glycerol dehydrogenase catalyzes the conversion of glycerol to glycerone.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?