• USA Home
  • G4884 - α-D-Galacturonic acid 1-phosphate lithium salt

EMAIL THIS PAGE TO A FRIEND
G4884 Sigma

α-D-Galacturonic acid 1-phosphate lithium salt

  • Empirical Formula (Hill Notation) C6H11O10P · xLi+

  • Molecular Weight 274.12 (free acid basis)

  •  PubChem Substance ID 24895190

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates G, Monosaccharide More...
storage temp.   −20°C

Description

General description

α-D-Galacturonic acid was found to be a component of Lipid A isolated from Mesorhizobium huakuii.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?