• USA Home
  • G4890 - Globulins Cohn fraction II, III from rat

EMAIL THIS PAGE TO A FRIEND
G4890 Sigma

Globulins Cohn fraction II, III from rat

lyophilized powder

Purchase

Description

Other Notes

Primarily β- and γ-globulins

Physical form

Lyophilized powder containing NaCl

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Plasma Derived Proteins and Enzymes

Despite their complexity, blood and plasma are abundant biological resources for the discovery of drug targets and biomarkers for human disease. It is estimated that plasma may contain as many as 40,...
BioFiles 2006, 1.5, 2.
Keywords: Applications, Gas chromatography, Type

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?