• USA Home
  • G4899 - Anti-Guanosine 3′,5′-Cyclic Monophosphate (cGMP) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
G4899 Sigma

Anti-Guanosine 3′,5′-Cyclic Monophosphate (cGMP) antibody produced in rabbit

whole antiserum, lyophilized powder

Synonym: Anti-cGMP

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies and Kits to Cyclic Nucleotides, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology,
biological source   rabbit
antibody form   whole antiserum
clone   polyclonal
form   lyophilized powder
application(s)   radioimmunoassay: suitable
conjugate   unconjugated
storage temp.   2-8°C

Description

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Immunogen

Cyclic GMP-2′-O-HS-BSA.

Packaging

100 test in glass bottle

Reconstitution

Reconstitute with 1 mL 0.1% BSA in distilled water. Store the reconstituted stock at −20 °C.

Specificity

The product is specific for cGMP and does not cross-react with 5′-AMP, 5′-ADP, 5′-ATP, cAMP, Guanosine, 5′-GMP, 5′-GDP and 5′-GTP. The antiserum is characterized utilizing the ethanol precipitation radioimmunoassay (RIA) protocol.
Working dilution is at least 1:10 of reconstituted antiserum (1 mL).

Application

Anti-Guanosine 3′,5′-Cyclic Monophosphate (cGMP) antibody produced in rabbit is suitable for the measurement of cGMP using ELISA and western blot analysis and for the immunolocalization of cGMP-containing cells in the preoptic region (PO). The antibody is also suitable for use in an ethanol precipitation radioimmunoassay (RIA) procedure.

Biochem/physiol Actions

cGMP acts as a secondary messenger in regards to hormone action like cAMP. Some cellular functions may be controlled bi-directionally by cGMP and cAMP. They may be stimulated by cGMP and suppressed by cAMP and vice versa. cGMP mediates smooth muscle relaxation.

General description

Cyclic GMP is present in most tissues at concentrations at least 10-fold and occasionally 100-fold lower than those of cAMP. However, a few tissues (brain, testes, and lung) are relatively rich in cGMP and low in cAMP. cGMP levels in plasma and urine are approximately half those of cAMP.

Price and Availability


Detect Your Unseen Protein
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer - Over 80,000 Products

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?