• USA Home
  • G5027 - L-Glutamic Dehydrogenase (NADP) from Candida utilis

EMAIL THIS PAGE TO A FRIEND
G5027 Sigma

L-Glutamic Dehydrogenase (NADP) from Candida utilis

lyophilized powder, 50-200 units/mg protein

Synonym: L-Glutamate:NADP+ oxidoreductase (deaminating)

Purchase

Description

Other Notes

Note: Do not confuse with non-specific L-GLDH, EC 1.4.1.3.

Unit Definition

One unit will reduce 1.0 μmole of α-ketoglutarate to L-glutamate per min at pH 8.3 at 30 °C in the presence of ammonium ions and NADPH.

Physical form

Lyophilized powder containing potassium phosphate buffer salt

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?