• USA Home
  • G5143 - Anti-Guinea Pig Serum antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
G5143 Sigma

Anti-Guinea Pig Serum antibody produced in rabbit

whole antiserum

Synonym: Anti-Serum, guinea pig

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies to Animal Plasma Proteins, GR-GZ, Primary Antibodies,
biological source   rabbit
antibody form   whole antiserum
clone   polyclonal
contains   15 mM sodium azide
species reactivity   guinea pig
application(s)   immunoelectrophoresis: suitable
conjugate   unconjugated
shipped in   dry ice
storage temp.   −20°C

Description

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?