• USA Home
  • G5382 - Glutaminase from Escherichia coli

EMAIL THIS PAGE TO A FRIEND
G5382 Sigma

Glutaminase from Escherichia coli

Grade II, lyophilized powder, 5-15 units/mg protein

Synonym: L-Glutamine amidohydrolase

Purchase

Properties

Related Categories 3.5.x.x Acting on C-N other than peptides, 3.x.x.x Hydrolases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
type   Grade II
form   lyophilized powder
composition   Protein, ~50% biuret
cation traces   NH4+: ≤1 μg/unit
storage temp.   −20°C

Description

Application

Suitable for use in determining glutamine by measuring ammonia formation. Contains glutamic decarboxylase which destroys some of the glutamate as it is formed.

Unit Definition

One unit will deaminate 1.0 μmole of L-glutamine per min at pH 4.9 at 37 °C.

Physical form

Lyophilized powder containing buffer salts

Biochem/physiol Actions

Glutaminase catalyzes the conversion of glutamine to glutamate.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
MA2275050
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?