EMAIL THIS PAGE TO A FRIEND
G5504 Sigma

Gly-His-Gly

  • CAS Number 7758-33-0

  • Empirical Formula (Hill Notation) C10H15N5O4

  • Molecular Weight 269.26

  •  PubChem Substance ID 24895213

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides and Tripeptides, Peptides, Tripeptides
InChI Key   ADZGCWWDPFDHCY-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?