• USA Home
  • G5506 - Glucose-6-phosphate Dehydrogenase–Agarose from baker's yeast (S. cerevisiae)

EMAIL THIS PAGE TO A FRIEND
G5506 Sigma

Glucose-6-phosphate Dehydrogenase–Agarose from baker's yeast (S. cerevisiae)

ammonium sulfate suspension, 1,000-1,750 units/g agarose, 30 °C

Purchase

Description

Unit Definition

One unit will oxidize 1.0 μmole of D-glucose 6-phosphate to 6-phospho-D-gluconate per min in the presence of NADP at pH 7.4 at 30 °C.

Physical form

Suspension in 2.0 M (NH4)2SO4, pH 7.0, containing 0.02% thimerosal

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Articles

Ammonium Sulfate Suspensions - Technical Note

Note that for an ammonium sulfate suspension, most of the enzyme will be in solid form. It is likely that only negligible amounts of enzyme will be in the ammonium sulfate solution.
Keywords: Enzyme activity

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?