• USA Home
  • G5524 - Glutathione S-Transferase Isozyme A1-1 human

EMAIL THIS PAGE TO A FRIEND
G5524 Sigma

Glutathione S-Transferase Isozyme A1-1 human

≥95% (SDS-PAGE), recombinant, expressed in E. coli overproducing strain, buffered aqueous glycerol solution, >50 U/mg

Synonym: GST A1-1

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes, Cell Signaling and Neuroscience,
recombinant   expressed in E. coli overproducing strain
assay   ≥95% (SDS-PAGE)
form   buffered aqueous glycerol solution
mol wt   mol wt ~45 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... GSTA1(2938)

Description

Biochem/physiol Actions

Glutathione S-transferases are a family of proteins that catalyze the conjugation of reduced glutathione with a variety of hydrophobic compounds containing an electrophilic center. Their major biological function is believed to be defense against electrophilic chemical species, many of which are formed by cellular oxidative reactions catalyzed by cytochrome P450 and other oxidases. The different specificities of the three GST isozymes are demonstrated by determining the I<SUB>50</SUB> values obtained using the inhibitor S-hexylglutathione in the standard 1-chloro-2,4-dinitrobenze<WBR>ne assay. I<SUB>50</SUB> (A1-1) < I<SUB>50</SUB> (M1-1) < I<SUB>50</SUB> (P1-1).

General description

GST A1-1 is found in only a few body tissues, including kidney, intestine, lung, and liver.

Unit Definition

One unit will catalyze the conjugation of 1.0 μmole of 1-chloro-2,4-dinitrobenzene and reduced glutathione per min at pH 6.5 at 25 °C.

Physical form

Solution in 50 mM Tris-HCl, pH 7.5, 50 mM NaCl, 1 mM DTT, 1 mM EDTA, and 50% glycerol.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

CS0410 Glutathione-S-Transferase (GST) Assay Kit, sufficient for 500 multiwell tests

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?