EMAIL THIS PAGE TO A FRIEND
G5629 Sigma

Gly-Leu-Tyr

Synonym: Glycyl-L-leucyl-L-tyrosine

  • CAS Number 4306-24-5

  • Empirical Formula (Hill Notation) C17H25N3O5

  • Molecular Weight 351.40

  •  Beilstein Registry Number 2179546

  •  EC Number 224-319-1

  •  MDL number MFCD00043036

Purchase

Properties

InChI Key   AFWYPMDMDYCKMD-KBPBESRZSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?