• USA Home
  • G5750 - α-D-Glucose 1,6-bisphosphate potassium salt hydrate

EMAIL THIS PAGE TO A FRIEND
G5750 Sigma

α-D-Glucose 1,6-bisphosphate potassium salt hydrate

≥93%

Synonym: α-D-Glucose 1,6-diphosphate potassium salt hydrate

  • CAS Number 91183-87-8

  • Empirical Formula (Hill Notation) C6H10K4O12P2 · xH2O

  • Molecular Weight 492.48 (anhydrous basis)

  •  MDL number MFCD00150469

  •  PubChem Substance ID 24895224

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates G, Monosaccharide More...
InChI Key   OVRXVCAVDXBPQT-QMKHLHGBSA-J
assay   ≥93%
storage temp.   −20°C

Description

Preparation Note

Enzymatically prepared.

Application

α-D-Glucose 1,6-bisphosphate is used in the identification, differentiation and characterization of α-phosphoglucomutase(s), β-phosphoglucomutase(s) and α- and β-hexoglucomutase(s).

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?