• USA Home
  • G6004 - Glutathione Reductase from wheat germ

EMAIL THIS PAGE TO A FRIEND
G6004 Sigma

Glutathione Reductase from wheat germ

Type II, ≥0.08 units/mg protein

Synonym: GR, NAD(P)H: oxidized-glutathione oxidoreductase

Purchase

Description

Biochem/physiol Actions

A crucial flavoenzyme in the antioxidant defense system. Reduced glutathione (GSH) is used by glutathione peroxidase to detoxify hydrogen peroxide and in the process is converted to oxidized glutathione (GSSG). The GSSG is then recycled back to GSH by glutathione reductase (GR) using NADPH that is then converted to NADP+. The regenerated GSH is then available to detoxify more hydrogen peroxide. The enzyme uses FAD as a cofactor. GR and glutathione peroxidase may inhibit lipid peroxidation by functioning as antioxidant enzymes in sperm. Glutathione reductase shares a structural motif with a number of other proteins including aspartyl proteases, citrate synthase, EF hands, hemoglobins, lipocalins, and α/β hydrolases. GR is stimulated by melatonin and is reportedly irreversibly inhibited by a number of oxygen radical generating systems.

Quality

Crude

Unit Definition

One unit will reduce 1.0 μmole of oxidized glutathione per min at pH 7.6 at 25 °C. (Protein determined by biuret).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

GRSA Glutathione Reductase Assay Kit

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?