• USA Home
  • G6755 - α-Glycerophosphate Dehydrogenase-Triosephosphate Isomerase from rabbit muscle

EMAIL THIS PAGE TO A FRIEND
G6755 Sigma

α-Glycerophosphate Dehydrogenase-Triosephosphate Isomerase from rabbit muscle

Type X, lyophilized powder, GDH ≥125 units/mg protein, TPI ≥1,250 units/mg protein

Synonym: α-GDH-TPI

Purchase

Properties

Related Categories 1.1.x.x Acting on hydroxyl groups, 1.x.x.x Oxidoreductases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
type   Type X
form   lyophilized powder
composition   Protein, ~60% modified Warburg-Christian
storage temp.   −20°C

Description

Packaging

(Sold on the basis of α-GDH units.)

Unit Definition

(α-GDH) One unit will convert 1.0 μmole of dihydroxyacetone phosphate to α-glycerophosphate per min at pH 7.4 at 25 °C.

(TPI) One unit will convert 1.0 μmole of D-glyceraldehyde 3-phosphate to dihydroxyacetone phosphate per min at pH 7.6 at 25 °C.

Physical form

Lyophilized sulfate-free powder containing buffer salts as citrate and EDTA

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Protocols

Enzymatic Assay of Fructose-6-Phosphate Kinase Pyrophosphate Dependent

To standardize a procedure for the enzymatic assay of Fructose -6- Phosphate Kinase (Pyrophosphate Dependent) from Propionibacterium Freudenreichii (Shermanii).
Keywords: Atomic layer deposition, Extinction coefficient

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

G1881 α-Glycerophosphate Dehydrogenase-Triosephosphate Isomerase from rabbit muscle, Type III, ammonium sulfate suspension, TPI 750-2000 units/mg protein, GDH 75-200 units/mg protein (biuret)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?