• USA Home
  • G6763 - Globulins Cohn fraction III human

EMAIL THIS PAGE TO A FRIEND
G6763 Sigma

Globulins Cohn fraction III human

Purchase

Properties

impurities   HIV and HBsAg, source material tested negative
storage temp.   2-8°C

Description

Other Notes

Predominantly γ- and β-globulins. Contains plasminogen activity.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

G2388 Globulins Cohn fraction II, III human, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?