• USA Home
  • G6901 - Monosialoganglioside-GM3-pyrene labeled

EMAIL THIS PAGE TO A FRIEND
G6901 Sigma

Monosialoganglioside-GM3-pyrene labeled

from bovine brain, ~95%, vacuum-dried powder

Synonym: GM3-pyrene

Purchase

Properties

Related Categories Acidic Glycosphingolipids, Biochemicals and Reagents, Gangliosides, Lipids, Sphingolipids More...
biological source   from bovine brain
assay   ~95%
form   vacuum-dried powder
storage temp.   −20°C

Description

Analysis Note

Labeled GM3 gives blue fluorescence with activation at 366 nm

Other Notes

Gangliosides are major constituents of neuronal cell membranes and endoplasmic reticulum. They contain a sialated polysaccharide chain linked to ceramide through a β-glycosidic linkage. For classification of gangliosides see Svennerholm, L., et al. (eds.), Structure and Function of Gangliosides, New York, Plenum, 1980.

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?