• USA Home
  • G6906 - β-Glucosidase from Caldocellum saccharolyticum

EMAIL THIS PAGE TO A FRIEND
G6906 Sigma

β-Glucosidase from Caldocellum saccharolyticum

recombinant, expressed in E. coli, lyophilized powder, ≥300 units/g solid

Synonym: β-D-Glucoside glucohydrolase

Purchase

Description

Biochem/physiol Actions

Hydrolysis of terminal, non-reducing β-D-glucose residues with release of β-D-glucose

Other Notes

Thermostable enzyme

Unit Definition

One unit will liberate 1.0 μmole of glucose from salicin per min at pH5.0 at 37°C.

Price and Availability


Amplified Detection

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
1
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

G4511 β-Glucosidase from almonds, lyophilized powder, 10-30 units/mg solid

inhibitor

Product #

Description

Add to Cart

C5913 4-Chloromercuribenzoic acid, cysteine active site modifier
C5424 Conduritol B epoxide
C3784 Castanospermine, ≥94% (GC), BioUltra, from Castanospermum australe seeds

substrate

Product #

Description

Add to Cart

S0625 D-(−)-Salicin, ≥99% (GC)
M3633 4-Methylumbelliferyl β-D-glucopyranoside, β-glucosidase substrate
A3582 3-Acetylumbelliferyl β-D-glucopyranoside, ≥95%
B4527 5-Bromo-4-chloro-3-indolyl β-D-glucopyranoside, ≥97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?