• USA Home
  • G7152 - Gel scoop UV-transparent acrylic
EMAIL THIS PAGE TO A FRIEND
G7152 Sigma
Gel scoop UV-transparent acrylic
size 20 cm × 20 cm
Purchase
Properties
Related Categories Electrophoresis Supplies, Gel Handling, Labware More...
size   20 cm × 20 cm
Description
General description
Constructed of 1/8 in. acrylic for easy transfer of polyacrylamide and agarose gels between electrophoresis unit, stain/destain tray, and transilluminator. Design includes offset handle, a beveled edge to simplify lifting gel, and upturned sides to keep the gel on the blade.
Dimensions are for the flat part of the scoop.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?