• USA Home
  • G7272 - Gradient-forming devices
EMAIL THIS PAGE TO A FRIEND
G7272 Sigma
Gradient-forming devices
total capacity 300 mL
Purchase
Properties
Related Categories Glass Chromatography Columns, Glassware, Labware, Sigma
total capacity   300 mL
Description
General description
Constructed of two equal-diameter heavy-walled acrylic cylinders secured to an acrylic base. PTFE stopcock between cylinders is closed for filling, then opened. Exit port from mixing chamber is controlled by addition of hose and pinch clamp. Geometry permits use of magnetic stirring bar in mixing chamber.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?