• USA Home
  • G7390 - Gallic acid lauryl ester

EMAIL THIS PAGE TO A FRIEND
G7390 Sigma

Gallic acid lauryl ester

Synonym: Lauryl 3,4,5-trihydroxybenzoate

Purchase

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
1
RTECS 
DH9100000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?