• USA Home
  • G7510 - Guanylate Kinase from bovine brain

EMAIL THIS PAGE TO A FRIEND
G7510 Sigma

Guanylate Kinase from bovine brain

lyophilized powder, 10-40 units/mg protein

Synonym: ATP:[d]GMP phosphotransferase, Deoxyguanylate kinase

Purchase

Properties

Related Categories 2.7.x.x Phosphorus containing groups, 2.x.x.x Transferases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
form   lyophilized powder
composition   Protein, ~50% modified Warburg-Christian
foreign activity   Myokinase, may contain trace amount
storage temp.   −20°C

Description

Unit Definition

One unit will convert 1.0 μmole each of GMP and ATP to GDP and ADP per min at pH 7.5 at 30 °C.

Physical form

Lyophilized powder containing potassium phosphate buffer salts

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?