• USA Home
  • G7515 - γ-Globulins from cat

EMAIL THIS PAGE TO A FRIEND
G7515 Sigma

γ-Globulins from cat

99% (agarose gel electrophoresis)

Purchase

Properties

Related Categories Biochemicals and Reagents, Globulins, Plasma & Blood Proteins, Plasma, Blood, and Related Proteins and Reagents, Proteins and Derivatives More...
assay   99% (agarose gel electrophoresis)
impurities   2-4% NaCl
storage temp.   −20°C

Description

Preparation Note

Prepared from Cohn Fraction II, III

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Plasma Derived Proteins and Enzymes

Despite their complexity, blood and plasma are abundant biological resources for the discovery of drug targets and biomarkers for human disease. It is estimated that plasma may contain as many as 40,...
BioFiles 2006, 1.5, 2.
Keywords: Applications, Gas chromatography, Type

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?