EMAIL THIS PAGE TO A FRIEND
G7520 Sigma

D-(+)-Glucose

plant cell culture tested, BioReagent

Synonym: Dextrose

Purchase

Properties

Related Categories Cell Biology, Glycolysis, Glycolysis Metabolites, Metabolic Pathways, Metabolomics More...
grade   BioReagent
InChI Key   WQZGKKKJIJFFOK-DVKNGEFBSA-N
product line   BioReagent
form   powder
mp   150-152 °C(lit.)
solubility   H2O: soluble2g + 15 mL, clear, colorless
suitability   plant cell culture tested
Featured Industry   Agriculture
Gene Information   human ... PYGM(5837)

Description

Other Notes

Mixed anomers

Packaging

1 kg in poly bottle

Application

D-(+)-Glucose (G7520) is plant cell culture tested (15 mg/ml) and is appropriate for use in plant cell culture experiments.

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
LZ6600000
Protocols & Articles

Related Content

Plant Tissue Culture Products | Agriculture

Our combination of quality products, services and global business capabilities help you to maintain reagent consistency while ensuring suitability. Sigma-Aldrich allows you to standardize reagents ac...
Keywords: Antibiotics, Cell culture, Culture media, Gas chromatography, High performance liquid chromatography, Mass spectrometry, Vitamins, transformation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?