• USA Home
  • G7529 - Glycoprotein IIb/IIIa for Dupont

EMAIL THIS PAGE TO A FRIEND
G7529 Sigma

Glycoprotein IIb/IIIa for Dupont

Purchase

Properties

storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?