• USA Home
  • G7541 - Genome Visualization by Classic Methods in Light Microscopy
EMAIL THIS PAGE TO A FRIEND
G7541 Sigma
Genome Visualization by Classic Methods in Light Microscopy
  •  ISBN-10 0-8493-0043-6
  •  ISBN-13 978-0-8493-0043-1
Purchase
Properties
Related Categories Biotechnology Books, Books and Software, Labware More...
publication info   J.M. Exbrayat, CRC Press, 2001, 216 pp., hard cover
mfr. no.   CRC Press
Description
General description
In a two column format, the author investigates the classic techniques employed in the visualization of nucleic acids by light microscopy. The book discusses the chemical structure of nucleic acids, general principles of staining, nuclear dyes, and histochemical methods. Each chapter features different methods of staining, along with chemical principles and protocols. It also contains a color section with examples of observations.
Table of Contents
Intro
Chemical structure of nucleic acids
Chemical principles for nucleic acids visualization
Fixation
Staining by nuclear dyes
Histochemistry of nucleic acids
Fluorescent dyes
Controls
Conclusions
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?