• USA Home
  • G7667 - Drawer liners, Groovy Pad
EMAIL THIS PAGE TO A FRIEND
G7667 Sigma
Drawer liners, Groovy Pad
size Long, L × W 18 in. × 24 in.
Purchase
Properties
Related Categories Drawer organzers, Labware, Storage More...
size   Long
L × W   18 in. × 24 in.
Description
General description
Groovy Pad comes in six sizes to fit drawers or benchtop area. Made from woven nylon, the ribbed material keeps thermometers, columns and other long, round objects from rolling when drawers are opened or closed
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?