EMAIL THIS PAGE TO A FRIEND
G7791 Sigma

GW8510

solid

Synonym: 4-{[(7-Oxo-6,7-dihydro-8H-[1,3]thiazolo[5,4-e]indol-8-ylidene)methyl]amino}-N-(2-pyridyl)benzenesulfonamide

Purchase

Description

Biochem/physiol Actions

GW8510 is an inhibitor of cyclin kinase 2 (CDK2). It prevents and/or reduces the side effects of chemotherapy (alopecia, plantar-palmar syndrome and mucositis).

Packaging

1, 5 mg in glass bottle

Legal Information

Sold for research purposes under agreement from Glaxo­Smith­Kline

Price and Availability


Better Together

Discover your Target

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?