• USA Home
  • G8001 - Glycine ethyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
G8001 Sigma

Glycine ethyl ester hydrochloride

≥98.5%

Synonym: Ethyl glycinate hydrochloride

  • CAS Number 623-33-6

  • Linear Formula NH2CH2COOC2H5 · HCl

  • Molecular Weight 139.58

  •  Beilstein Registry Number 3594034

  •  EC Number 210-787-4

  •  MDL number MFCD00012871

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories A - H, Amino Acids, Amino Acids & Derivatives, Biochemicals and Reagents, Chemical Biology,
InChI Key   TXTWXQXDMWILOF-UHFFFAOYSA-N
assay   ≥98.5%
mp   145-146 °C(lit.)

Description

Other Notes

This product has been replaced by G6503-ALDRICH | Glycine ethyl ester hydrochloride 99%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
MC0525000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

G6503 Glycine ethyl ester hydrochloride, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?