• USA Home
  • G8002 - Guanosine 2′- & 3′-monophosphate Mixed Isomers sodium salt

EMAIL THIS PAGE TO A FRIEND
G8002 Sigma

Guanosine 2′- & 3′-monophosphate Mixed Isomers sodium salt

Synonym: Guanylic acid

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleotides, Oligonucleotides, Nucleotide Analogs More...
storage temp.   −20°C

Description

Linkage

Do not confuse with guanosine 2′:3′-cyclic monophosphate.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?