• USA Home
  • G8040 - γ-Glutamyltranspeptidase from bovine kidney

EMAIL THIS PAGE TO A FRIEND
G8040 Sigma

γ-Glutamyltranspeptidase from bovine kidney

Type I, 2.0-10 unit/mg solid

Synonym: γ-GT, γ-Glutamyltransferase, (5-Glutamyl)peptide:amino-acid 5-glutamyltransferase, GGTP

Purchase

Description

Quality

Crude

Unit Definition

One unit will liberate 1.0 μmole of p-nitroaniline from L-γ-glutamyl-p-nitroanilide per min at pH 8.5 at 25°C.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

inhibitor

Product #

Description

Add to Cart

I2512 Sodium iodoacetate, ≥98%

replaced by

Product #

Description

Add to Cart

G2262 γ-Glutamyltranspeptidase from porcine kidney, Type IV, 2.0-6.0 units/mg solid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?