• USA Home
  • G8145 - Guanosine 5′-monophosphate–Agarose

EMAIL THIS PAGE TO A FRIEND
G8145 Sigma

Guanosine 5′-monophosphate–Agarose

saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Molecular Biology, Nucleotide/Coenzyme Resins, Protein Chromatography, Proteomics More...
form   saline suspension
extent of labeling   2-4 μmol per mL
matrix   4% beaded agarose
matrix activation   cyanogen bromide
matrix attachment   ribose hydroxyls
matrix spacer   11 atoms (adipic acid dihydrazide)
storage temp.   2-8°C

Description

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Price and Availability


Amplified Detection

Amicon Pro Purification System
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?