• USA Home
  • G8165 - Galanin (1-13)-Neuropeptide Y (25-36) Amide

EMAIL THIS PAGE TO A FRIEND
G8165 Sigma

Galanin (1-13)-Neuropeptide Y (25-36) Amide

≥90%

Synonym: m32

  • Empirical Formula (Hill Notation) C136H209N41O34

  • Molecular Weight 2962.37

  •  MDL number MFCD00798702

Purchase

Description

Amino Acid Sequence

Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH2

Biochem/physiol Actions

This chimeric peptide is a high affinity galanin receptor antagonist (IC50=0.1 nM in rat hypothalamus membranes and Kd=0.01 nm in rat spinal cord membranes). This peptide is also recognized by neuropeptide Y receptors (IC50=0.25 μM in rat cerebral cortex membranes) and is a Y2 agonist in rat vas deferens.

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?