• USA Home
  • G8262 - Guanosine 5′-triphosphate, periodate oxidized sodium salt

EMAIL THIS PAGE TO A FRIEND
G8262 Sigma

Guanosine 5′-triphosphate, periodate oxidized sodium salt

85-90%

Synonym: Guanosine 5′-triphosphate-2′,3′-dialdehyde

Purchase

Properties

Related Categories Aldehydes, Biochemicals and Reagents, Building Blocks, C10-C12, Carbonyl Compounds,
InChI Key   XLIUTIYREXVACK-KGZKBUQUSA-M
assay   85-90%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?