• USA Home
  • G8271 - β-Glucuronidase from Escherichia coli

EMAIL THIS PAGE TO A FRIEND
G8271 Sigma

β-Glucuronidase from Escherichia coli

Preweighed vials of G7646, vial of ≥1000 units

Synonym: β-D-Glucuronide glucuronosohydrolase

Purchase

Description

Quality

Does not contain added buffer.

Unit Definition

One Sigma or modified Fishman unit will liberate 1.0 μg of phenolphthalein from phenolphthalein glucuronide per hr at 37 °C at the pH 6.8 (30 min assay).

Application

β-glucuronidase is used as a reporter gene in GUS assays to monitor gene expression.

Biochem/physiol Actions

β-glucuronidase catalyzes the breakdown of complex carbohydrates. In humans it converts conjugated bilirubin into the unconjugated form, making bilirubin suitable for reabsorption.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
LZ8990000
Protocols & Articles

Related Content

Analytical Enzyme Beta Glucuronidase

Our β-Glucuronidases are routinely used for the enzymatic hydrolysis of glucuronides from urine1,2, plasma3,4, and other fluids5 prior to analysis by enzyme immunoassay, mass spectrometry, gas chroma...
Keywords: Bacterial conjugations, Cancer, Chromatography, Gas chromatography, High performance liquid chromatography, Immunoassay, Mass spectrometry, PAGE, Purification

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

G7646 β-Glucuronidase from Escherichia coli, Type VII-A, lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)

substrate

Product #

Description

Add to Cart

73677 4-Nitrophenyl β-D-glucuronide, ≥99.0% (TLC)
B8174 5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt, tablet
B4532 5-Bromo-6-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?