• USA Home
  • G8386 - Glutathione S-Transferase from rat liver

EMAIL THIS PAGE TO A FRIEND
G8386 Sigma

Glutathione S-Transferase from rat liver

lyophilized powder, ≥15 units/mg protein

Synonym: Glutathione S-alkenetransferase, Glutathione S-alkyltransferase, Glutathione S-aralkyltransferase, Glutathione S-aryltransferase, Glutathione S-epoxidetransferase, RX: Glutathione R-transferase

Purchase

Properties

Related Categories 2.5.x.x Alkyl or aryl groups other than methyl, 2.x.x.x Transferases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
form   lyophilized powder
composition   Protein, ~75%
storage temp.   −20°C
Gene Information   rat ... Gsta3(24421), Gstcd(310855)

Description

Analysis Note

Protein determined by biuret.
Purified and assayed by a modification of the method of Simons and Vander Jagt.
Enzymatic activities are based on the conjugation of reduced glutathione with a second substrate. The individual proteins generally have activity with more than one class of substrate.

Unit Definition

One unit will conjugate 1.0 μmole of 1-chloro-2,4-dinitrobenzene with reduced glutathione per min at pH 6.5 at 25°C.

Physical form

Lyophilized powder containing Tris, phosphate salts, reduced glutathione and EDTA

Biochem/physiol Actions

Glutathione S-transferases are a family of proteins that catalyze the conjugation of reduced glutathione with a variety of hydrophobic chemicals containing electrophilic centers.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

G6511 Glutathione S-Transferase from equine liver, lyophilized powder, ≥25 units/mg protein

inhibitor

Product #

Description

Add to Cart

T9034 Taurocholic acid sodium salt hydrate, BioXtra, ≥95% (TLC)
T4512 (±)-Taxifolin hydrate, ≥90% (HPLC)
E3520 Ebselen, cysteine modifier

related product

Product #

Description

Add to Cart

CS0410 Glutathione-S-Transferase (GST) Assay Kit, sufficient for 500 multiwell tests

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?