EMAIL THIS PAGE TO A FRIEND
G8389 Sigma

Glass wool

Synonym: Fiberglass

Purchase

Properties

mfr. no.   Corning

Description

Other Notes

Pyrex™ brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
LK3651000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?