• USA Home
  • G8635 - β-Glucuronidase, molluscan from Chlamys opercularis (scallops)

EMAIL THIS PAGE TO A FRIEND
G8635 Sigma

β-Glucuronidase, molluscan from Chlamys opercularis (scallops)

Type S-I, lyophilized powder, 5,000-10,000 units/g solid, pH 3.8 (30 min assay), secondary activity: ≤800 units/g solid sulfatase

Synonym: β-D-Glucuronide glucuronosohydrolase

Purchase

Properties

Related Categories 3.2.x.x Glycosidases, 3.x.x.x Hydrolases, Biochemicals and Reagents, Enzyme Class Index, Enzymes, Inhibitors, and Substrates More...
type   Type S-I
form   lyophilized powder
storage temp.   −20°C

Description

Quality

Many β-glucuronidases derived from mollusks also contain sulfatase activity.
Some preparations have been reported to contain acid phosphatase activity.

Unit Definition

One Sigma or modified Fishman unit will liberate 1.0μg of phenolphthalein from phenolphthalein glucuronide per hr at 37°C at the appropriate pH (30min assay). Sulfatase Unit Definition: One unit of sulfatase will hydrolyze 1.0μmole p-nitrocatechol sulfate per hr at pH5.0 at 37°C.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3
RTECS 
LZ8990000

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

G0751 β-Glucuronidase from Helix pomatia, Type H-1, partially purified powder, ≥300,000 units/g solid

substrate

Product #

Description

Add to Cart

N7006 4-Nitrophenyl β-D-glucopyranoside, ≥98% (TLC)
B8174 5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt, tablet
B4532 5-Bromo-6-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?