• USA Home
  • G8663 - Monosialoganglioside GM1 from bovine brain

EMAIL THIS PAGE TO A FRIEND
G8663 Sigma

Monosialoganglioside GM1 from bovine brain

Synonym: Ganglioside GM1, monosialo

Purchase

Properties

storage temp.   −20°C

Description

Amino Acid Sequence

Cer-Glc-Gal(NeuAc)-GalNAc-Gal

Application

Specific receptors for cholera toxin that accumulate in the brain in late infantile lipidosis.

General description

Gangliosides are major constituents of neuronal cell membranes and endoplasmic reticulum. They contain a sialated polysaccharide chain linked to ceramide through a β-glycosidic linkage. For classification of gangliosides see Svennerholm, L., et al. (eds.), Structure and Function of Gangliosides, New York, Plenum, 1980.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?