• USA Home
  • G8887 - Glucokinase from Bacillus stearothermophilus

EMAIL THIS PAGE TO A FRIEND
G8887 Sigma

Glucokinase from Bacillus stearothermophilus

lyophilized powder, ≥250 units/mg protein

Synonym: ATP:D-glucose-6-phosphotransferase

Purchase

Description

Unit Definition

One unit will phosphorylate 1.0 μmole of D-glucose to D-glucose 6-phosphate per min at pH 9.0 at 30 °C.

Physical form

Lyophilized powder containing Tris buffer salts

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?