• USA Home
  • G8909 - Anti-GroES antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
G8909 Sigma

Anti-GroES antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Cell Stress, Antibodies to Chaperonins and Heat Shock Proteins,
species reactivity   E. coli
application(s)   dot blot: 1:5,000 using 100 ng recombinant GroES/dot
  indirect ELISA: 1:15,000 using recombinant GroES protein
  western blot: 1:5,000 using a heat shocked E. coli cell extract
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   Escherichia coli ... groES(913836)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

recombinant GroES produced in E. coli coupled to KLH.

Physical form

Solution in in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Specificity

Does not cross-react with GroEL.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, half-mask respirator (US), multi-purpose combination respirator cartridge (US)
RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?