• USA Home
  • G9027 - L-Glutathione oxidized

EMAIL THIS PAGE TO A FRIEND
G9027 Sigma

L-Glutathione oxidized

~98%, lyophilized powder

Synonym: GSSG, Glutathiol

  • CAS Number 27025-41-8

  • Empirical Formula (Hill Notation) C20H32N6O12S2

  • Molecular Weight 612.63

  •  Beilstein Registry Number 1718700

  •  EC Number 248-170-7

  •  MDL number MFCD00063106

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Drug and Xenobiotic Metabolism, Multi-Drug Resistance and Drug Metabolism, Substrates, Inducers, Inhibitors More...
InChI Key   YPZRWBKMTBYPTK-BJDJZHNGSA-N
assay   ~98%
form   lyophilized powder
purified by   crystallization
impurities   Ethanol, free
solubility   H2O: soluble50 mg/mL, clear, colorless
storage temp.   2-8°C

Description

Amino Acid Sequence

Glu-Cys-Gly, Glu-Cys-Gly [Disulfide bridge: 2a-2b]

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

38185 Quantification kit for oxidized and reduced glutathione, 200 tests

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?