• USA Home
  • G9263 - Glyceraldehyde-3-phosphate Dehydrogenase from chicken muscle

EMAIL THIS PAGE TO A FRIEND
G9263 Sigma

Glyceraldehyde-3-phosphate Dehydrogenase from chicken muscle

lyophilized powder, ≥40 units/mg protein

Synonym: D-Glyceraldehyde 3-phosphate:NAD+ oxidoreductase (phosphorylating), GAPDH

Purchase

Properties

Related Categories 1.2.x.x Acting on aldehydes or oxo groups, 1.x.x.x Oxidoreductases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
form   lyophilized powder
composition   Protein, ≥85% biuret
foreign activity   3-Phosphoglyceric phosphokinase ≤0.1%
  Triose-phosphate isomerase ≤0.1%
storage temp.   −20°C

Description

Unit Definition

One unit will reduce 1.0 μmole of 3-phosphoglycerate to D-glyceraldehyde 3-phosphate per min in a coupled system with 3-phosphoglyceric phosphokinase at pH 7.6 at 25 °C.

Physical form

Lyophilized powder containing citrate buffer salts

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

inhibitor

Product #

Description

Add to Cart

I2512 Sodium iodoacetate, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?