EMAIL THIS PAGE TO A FRIEND
G9403 Sigma
GTPases
  •  ISBN-10 0-19-963744-X
  •  ISBN-13 978-0-19-963744-7
Purchase
Properties
Related Categories Biochemistry Books, Books and Software, Labware More...
publication info   A. Hall, ed., Oxford University Press, 2000, 360 pp., soft cover
mfr. no.   Oxford University Press
Description
General description
GTPases are proteins that act as molecular switches to control biochemical pathways within a cell. This book provides a complete guide to the properties and cellular roles of all the major families of GTPases, including the heterotrimeric G proteins and the small GTPases Ras, Rho, Rab, Arf and Ran. These proteins have caught the attention of those studying control of proliferation, differentiation, cell polarity, cell movement and vesicle and protein trafficking.
Table of Contents
Preface,
1. G proteins I: Gs and Gi,
2. G proteins II: Gq, G12, and Gz,
3. Ras,
4. Rho,
5. Rab,
6. Arf,
7. Ran,
8. GTPases in protein translocation and protein elongation
9. The structure and function of molecular switches in three dimensions,
10. GTPases targeted by bacterial toxins,
Index
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?