• USA Home
  • G9512 - Gly-Phe β-naphthylamide

EMAIL THIS PAGE TO A FRIEND
G9512 Sigma

Gly-Phe β-naphthylamide

crystalline

  • CAS Number 21438-66-4

  • Empirical Formula (Hill Notation) C21H21N3O2

  • Molecular Weight 347.41

  •  MDL number MFCD00021597

  •  PubChem Substance ID 24895376

Purchase

Description

Linkage

Similar to G 4255, but produced by Sigma.

Substrates

Substrate for cathepsin C

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?