• USA Home
  • G9541 - G Protein Antagonist

EMAIL THIS PAGE TO A FRIEND
G9541 Sigma

G Protein Antagonist

≥90% (HPLC)

  • CAS Number 143675-79-0

  • Empirical Formula (Hill Notation) C57H64N12O9S

  • Molecular Weight 1093.26

  •  MDL number MFCD03444578

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, G Protein Function, G Proteins and Cyclic Nucleotides, Reagents More...
assay   ≥90% (HPLC)
storage temp.   −20°C
Gene Information   human ... CHRM2(1129)

Description

Amino Acid Sequence

Glp-Gln-D-Trp-Phe-D-Trp-D-Trp-Met-NH2

Biochem/physiol Actions

Inhibits the activation of Gαi or Gαo by M2 muscarinic cholinergic receptor.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?