• USA Home
  • GASTRINS - Synthetic Gastrin Kit

EMAIL THIS PAGE TO A FRIEND
GASTRINS Sigma

Synthetic Gastrin Kit

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
shipped in   dry ice
storage temp.   −20°C

Description

Application

Contains a complete set of gastrin peptides including HG-17 in three forms. The complete hormones are 100% biologically active.

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters

Kit component only

Description

Product #

Add to Cart

[Leu15]-Gastrin I human, 100% (HPLC) 1 mg SDS    
[Nle15]-Gastrin 1, human, 99% (TLC) 1 mg SDS    
[Met15]-Gastrin 1 human, 99.58% (HPLC) 1 mg SDS    
[Leu15]-Gastrin 2-17, 99% 1 mg SDS    
Pentagastrin, solid 5 mg SDS    
Gly-Trp-Met-Asp-Phe-NH2 5 mg SDS    
Trp-Met-Asp-Phe-NH2 5 mg SDS    
Met-Asp-Phe-NH2 5 mg SDS    
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?